Learn More
Sigma Aldrich Fine Chemicals Biosciences Endothelin 1 97 HPLC powde
Catalog No. 502897590
Encompass
Endothelin-1 (ET-1) is synthesized and secreted by activated endothelial cells and is a peptide with potent vasoconstrictor activity. It is a marker for activated or dysfunctional endothelial cells. This peptide is one of the three isopeptides belonging to the endothelin family. It is also released by cardiomyocytes.
Catalog No. 50-289-7590 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. E776450UG
Endothelin-1 (ET-1) is synthesized and secreted by activated endothelial cells and is a peptide with potent vasoconstrictor activity. It is a marker for activated or dysfunctional endothelial cells. This peptide is one of the three isopeptides belonging to the endothelin family. It is also released by cardiomyocytes.