Learn More
Enzo Life Sciences Endothelin-1 ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900020A
Encompass_Preferred
Sensitive, rapid Endothelin-1 ELISA kit for cardiovascular and stress-related research. The Endothelin-1 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 2 hours. This ultra-sensitive kit lets you quantify as little as 0.41 pg/mL ET-1 in a variety of biological matrices with the option of overnight or a 1 hour primary incubation. Sensitivity: 0.41 pg/ml (range 0.78 - 100 pg/ml). Assay Time: 2 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human, Mouse, Rat, Rabbit, Porcine, Dog, Bovine. Contents: Microtiter plate, Antibody diluent, Antibody concentrate, Assay buffer 17, Wash buffer concentrate, Standard, TMB Substrate, Stop solution 2
Catalog No. 50-658-106