Learn More
Bachem Endothelin-1 (human, bovine, dog, mouse, porcine, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4048904.1000
Encompass_Preferred
Endothelin-1 (human, bovine, dog, mouse, porcine, rat) Trifluoroacetate, H-Cys-Ser-Cys-Ser-Ser-Leu-Met-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-His-Leu-Asp-Ile-Ile-Trp-OH, 117399-94-7, C109H159N25O32S5, Mr 2491.94, 1mg. ET-1 (human, bovine, dog, mouse, porcine, rat), Endothelin-1, human. Endothelin-1 (ET-1) is a potent vasoconstrictive 21-amino acid peptide produced by endothelia cells. It acts on endothelia cells and on the underlying smooth muscle cells as a modulator of vascular tone. ET-1 shows selectivity for the ET-A receptor.
Catalog No. 50-194-6779