Learn More
Medchemexpress LLC Endothelin 1 (swine, human) TFA | 394693-38-0 | 99.77% | 2605.92 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP0202A10MG
Encompass_Preferred
Endothelin 1 (swine, human) TFA is a synthetic peptide replicating the sequence of human and swine Endothelin 1. It acts as a potent endogenous vasoconstrictor by targeting ETA and ETB receptors. This product is intended for research use only.
  • Synthetic peptide with human and swine Endothelin 1 sequence
  • Potent endogenous vasoconstrictor
  • Acts through ETA and ETB receptors
  • For research use only
Catalog No. 50-004-25803