Learn More
Medchemexpress LLC Endothelin 1 (swine, human) TFA | 394693-38-0 | 99.8% | 2605.92 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP0202A5MG
Encompass_Preferred
Endothelin 1 (swine, human) TFA is a synthetic peptide that mimics the sequence of both human and swine Endothelin 1. This potent endogenous vasoconstrictor primarily functions by interacting with two types of receptors, ETA and ETB, playing a crucial role in vascular regulation.
  • Synthetic peptide
  • Mimics human and swine Endothelin 1 sequence
  • Potent endogenous vasoconstrictor
  • Acts on ETA and ETB receptors
  • Crucial for vascular regulation
Catalog No. 50-004-25791