Learn More
Medchemexpress LLC Endothelin 1 (swine, human) TFA | 394693-38-0 | 99.8% | 2605.92 | 500 UG
SDP
Supplier:  Medchemexpress LLC HYP0202A500UG
Encompass_Preferred
Endothelin 1 (swine, human) (TFA) is a synthetic peptide with the sequence of human and swine Endothelin 1. It is a potent endogenous vasoconstrictor and acts through two types of receptors, ETA and ETB. It is for research use only.
  • Synthetic peptide with the sequence of human and swine Endothelin 1
  • Potent endogenous vasoconstrictor
  • Acts through two types of receptors, ETA and ETB
  • For research use only
Catalog No. 50-004-26285