Learn More
Medchemexpress LLC HY-P0207A 500ug Medchemexpress, Endothelin-2 (49-69), human (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0207A500UG
Encompass_Preferred
Medchemexpress, HY-P0207A 500ug Endothelin-2 (49-69), human (TFA) CAS: Endothelin-2 (49-69), human (TFA) (Endothelin-2 (49-69) (human, canine) (TFA)) is a 21-amino acid vasoactive peptide that binds to G-protein-linked transmembrane receptors, ET-RA and ET-RB. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3030