Learn More
CPC Scientific H-Cys-Thr-Cys-Phe-Thr-Tyr-Lys-Asp-Lys-Glu-Cys-Val-Tyr-Tyr-Cys-His-Leu-Asp-Ile-Ile-Trp-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502109240
Encompass
ONE-LETTER SEQUENCE: CTCFTYKDKECVYYCHLDIIW (C1&C15 bridge, C3&C11 bridge)MOLECULAR FORMULA: C121H168N26O33S4MOLECULAR WEIGHT:2643.1STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [117399-93-6], RESEARCH AREA: CardiovascularREFERENCES: K. Nakajima et al., J. Cardiovasc. Pharmacol., 13, S8 (1989); A. Inoue, et al., PNAS, 86, 2863 (1989); M. Yanagisawa et al., PNAS, 85, 6964 (1988)
Catalog No. 50-210-9240 Supplier CPC Scientific Supplier No. ENDT005A
ONE-LETTER SEQUENCE: CTCFTYKDKECVYYCHLDIIW (C1&C15 bridge, C3&C11 bridge)MOLECULAR FORMULA: C121H168N26O33S4MOLECULAR WEIGHT:2643.1STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [117399-93-6], RESEARCH AREA: CardiovascularREFERENCES: K. Nakajima et al., J. Cardiovasc. Pharmacol., 13, S8 (1989); A. Inoue, et al., PNAS, 86, 2863 (1989); M. Yanagisawa et al., PNAS, 85, 6964 (1988)