Learn More
Medchemexpress LLC HY-P0204A 1mg Medchemexpress, Endothelin-3, human, mouse, rabbit, rat (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0204A1MG
Encompass_Preferred
Medchemexpress, HY-P0204A 1mg Endothelin-3, human, mouse, rabbit, rat (TFA) CAS: Endothelin-3, human, mouse, rabbit, rat TFA is a 21-amino acid vasoactive peptide that binds to G-protein-linked transmembrane receptors, ET-RA and ET-RB . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4049