Learn More
Cayman Chemical EndothelIn AChEFAb Conjugat
SDP
Catalog No. 502892290
Encompass_Preferred
An inhibitor of the Rho GEF 12-RhoA protein-protein interaction binds to the DH-PH domain of Rho GEF 12 in a cell-free assay (Kd 76 nM) inhibits serum-induced phosphorylation of MLC and FAK a marker of RhoA activity in NIH3T3 cells at 10 and 30 UM inhibits cell growth migration invasion and mammosphere formation in MCF-7 breast cancer cells from 5-30 UM
Catalog No. 50-289-2290 Supplier Cayman Chemical Supplier No. 483150500 DTN
An inhibitor of the Rho GEF 12-RhoA protein-protein interaction binds to the DH-PH domain of Rho GEF 12 in a cell-free assay (Kd 76 nM) inhibits serum-induced phosphorylation of MLC and FAK a marker of RhoA activity in NIH3T3 cells at 10 and 30 UM inhibits cell growth migration invasion and mammosphere formation in MCF-7 breast cancer cells from 5-30 UM