Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Endothermic Reactions
GREENER_CHOICESCI_ED

Catalog No. S96737 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S96737 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96737 Supplier Innovating Science™ Supplier No. IS7005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Materials for five demonstrations: Bags (5), 25 g Ammonium Nitrate, 1 Tbsp. Vermiculite, and instructions

Study the process of endothermic reactions.

  • Reaction is safely carried out in a plastic bag
  • Students can pass bag around to feel reaction

Compliance

Package conforms to 49 CFR 173.4

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Study the process of endothermic reactions
Includes 5 bags x 25 g ammonium nitrate un1942 + 1 tbsp vermiculite
Quantity 1
Dimensions (L x W x H) 4.25 x 4.625 x 5.5 in.
Class Size 15 Lab Groups
Product Type Endothermic Reactions Demonstration Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.