Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Endothermic Reactions
GREENER_CHOICESCI_ED

Catalog No. S96893 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S96893 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Acetyl Salicylic Acid/ Aspirin, which is known to the State of California to cause and reproductive harm.
Catalog No. S96893 Supplier Innovating Science™ Supplier No. IS2500
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Acetyl Salicylic Acid/ Aspirin, which is known to the State of California to cause and reproductive harm.

Includes

Sodium Bicarbonate/Citric Acid tablets (15); plastic cups (15); stirring sticks (15); 25g Ammonium Nitrate UN1942 (15); and 1 Tbsp. Vermiculite™ (15)

Requires

Thermometers, stopwatches (or a visible clock with second hand), 50mL graduated cylinder, and distilled water

Monitor change in temperature as the reaction occurs

Examine a commercial application that uses endothermic reactions: the instant cold pack. Enough materials for 15 groups.

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To examine a commercial application
Includes Sodium bicarbonate/citric acid tablets, 15 plastic cups, 15 stirring sticks, 15 bags containing 25g ammonium nitrate, 1 table spoon vermiculite, teacher's manual and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Product Type Endothermic Reaction Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.