Learn More
Bioworld ENDOTOXIN AFFISORBENTPOLY
Catalog No. 50197903
Encompass
ENDOTOXIN AFFISORBENT (POLYMYXIN B SEPAROPORE (AGAROSE)): bioWorld's Endotoxin Affisorbentconsists of immobilized polymyxin B on Separopore 6B-CL (6% crosslinked agarose beads). The polymyxins are a family of antibiotics that contain a c...
Catalog No. 50-197-903 Supplier Bioworld Supplier No. 201810921
ENDOTOXIN AFFISORBENT (POLYMYXIN B SEPAROPORE (AGAROSE)): bioWorld's Endotoxin Affisorbentconsists of immobilized polymyxin B on Separopore 6B-CL (6% crosslinked agarose beads). The polymyxins are a family of antibiotics that contain a c...