Learn More
Bioworld ENDOTOXIN AFFISORBENTPOLY
Supplier:  Bioworld 201810922
Encompass
ENDOTOXIN AFFISORBENT (POLYMYXIN B SEPAROPORE (AGAROSE)): bioWorld's Endotoxin Affisorbentconsists of immobilized polymyxin B on Separopore 6B-CL (6% crosslinked agarose beads). The polymyxins are a family of antibiotics that contain a c...
Catalog No. 50-197-904