Learn More
Bioworld ENDOTOXIN AFFISORBENTPOLY
Catalog No. 50197651
Encompass
ENDOTOXIN AFFISORBENT (POLYMYXIN B SEPAROPORE): bioWorld's Endotoxin Affisorbentconsists of immobilized polymyxin B on Separopore 6B-CL (6% crosslinked agarose beads). The polymixins are a family of antibiotics that contain a cationic cy...
Catalog No. 50-197-651 Supplier Bioworld Supplier No. 201700321
ENDOTOXIN AFFISORBENT (POLYMYXIN B SEPAROPORE): bioWorld's Endotoxin Affisorbentconsists of immobilized polymyxin B on Separopore 6B-CL (6% crosslinked agarose beads). The polymixins are a family of antibiotics that contain a cationic cy...