Learn More
Lonza Walkersville Control Standard Endotoxin
Catalog No. NC1577414
Encompass
This control standard endotoxin is used in combination with Kinetic-QCL Bulk Kinetic Chromogenic LAL Assay, K50-643L and K50-643U. It is available as a made-to-order product.Content:25 vials
Catalog No. NC1577414 Supplier Lonza Walkersville Supplier No. E50643L
This control standard endotoxin is used in combination with Kinetic-QCL Bulk Kinetic Chromogenic LAL Assay, K50-643L and K50-643U. It is available as a made-to-order product.Content:25 vials