Learn More
Charles River Laboratories ENDOTOXIN-FREE 96WL MCRO 50/CS
Catalog No. NC9217517
Encompass
96-Well Endosafe Polystyrene Plates (Individually wrapped) Certified to 0.005 EU/m
Catalog No. NC9217517 Supplier Charles River Laboratories Supplier No. M9005
96-Well Endosafe Polystyrene Plates (Individually wrapped) Certified to 0.005 EU/m