Learn More
Genscript Corporation ToxinSensor™ Endotoxin-free Pipette Tips (1 ml, Blue)
Catalog No. 50274656
Encompass
GenScript innovating Endotoxin-free pipette tips are designed for compatible use with most common pipetts from different manufacturers, which brings more flexibility. All tips are available in 1 version (1 ml) for laboratory routines. In addition, the tip can be used to dilute or aliquot sample with a low endotoxin level requirement (<0.005 EU/ml).
GenScript Endotoxin-free tips are provided in encapsulation with pyrogen-free plastic bags, which sufficiently inhibits the introduction of any contamination, such as RNase/DNase.
Catalog No. 50-274-656 Supplier Genscript Corporation Supplier No. M01063
GenScript innovating Endotoxin-free pipette tips are designed for compatible use with most common pipetts from different manufacturers, which brings more flexibility. All tips are available in 1 version (1 ml) for laboratory routines. In addition, the tip can be used to dilute or aliquot sample with a low endotoxin level requirement (<0.005 EU/ml).
GenScript Endotoxin-free tips are provided in encapsulation with pyrogen-free plastic bags, which sufficiently inhibits the introduction of any contamination, such as RNase/DNase.