Learn More
Cytiva Acrodisc unit with Mustang E membrane, 0.2 µm, 25 mm, non-sterile (10/pkg (blister packs))
Supplier:  Cytiva MSTG25E3
Encompass
Disposable Membrane Chromatography units are designed for the rapid selection of appropriate chemistry and method development. The Mustang membrane chromatography starter kit includes both strong anion and cation exchange membrane chromatography Acrodisc units, as well as specific endotoxin removal Acrodisc units. These allow both the rapid selection of the appropriate chemistry for your application, and the opportunity to quickly screen for optimal process parameters to enhance and simplify your process method development. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC0328470