Learn More
Bioworld Endotoxin Removal Kit (Mini) - For DNA, 20 Prep(s)
Catalog No. 50196963
Encompass_Preferred
For the rapid removal of endotoxins from previously purified DNA Reduce endotoxin levels to 0.1 EU/µg DNA or less Effectively remove endotoxins in as little as 20 minutes Remove endotoxins from up to 40 µg of DNA Rapid spin-column formatFEATURES AND BENEFITSFast and easy processing - Rapid spin-column format allows for the processing of multiple samples in 20 minutes Endotoxin-free plasmid DNA- Using Norgen’s Endotoxin Removal Kit (Mini), endotoxin levels are reduced to less than or equal to 0.1 EU/µg plasmid DNA High recovery of input DNA - Average DNA recovery is > 90% of the input amount Recovered DNA is suitable for downstream applications - Purified DNA is fully compatible with sequencing, cloning and transfections
Catalog No. 50-196-963 Supplier Bioworld Supplier No. 107601061
For the rapid removal of endotoxins from previously purified DNA Reduce endotoxin levels to 0.1 EU/µg DNA or less Effectively remove endotoxins in as little as 20 minutes Remove endotoxins from up to 40 µg of DNA Rapid spin-column formatFEATURES AND BENEFITSFast and easy processing - Rapid spin-column format allows for the processing of multiple samples in 20 minutes Endotoxin-free plasmid DNA- Using Norgen’s Endotoxin Removal Kit (Mini), endotoxin levels are reduced to less than or equal to 0.1 EU/µg plasmid DNA High recovery of input DNA - Average DNA recovery is > 90% of the input amount Recovered DNA is suitable for downstream applications - Purified DNA is fully compatible with sequencing, cloning and transfections