Learn More
Med Vet International Endotracheal Tube Adapter 12.0mm, J0157T
Catalog No. 5000583159
Encompass_Preferred
Portex Endotracheal Adaptor The full range of small animal endotracheal tubes is covered by the twelve adaptors. The adaptors may be sterilized by conventional methods. The compact Swivel Connector may be rotated a full 360 degrees affording maximum positioning and flexibility. For use on all endotracheal tubes.
Catalog No. 50-005-83159 Supplier Med Vet International Supplier No. JOR157T
Portex Endotracheal Adaptor The full range of small animal endotracheal tubes is covered by the twelve adaptors. The adaptors may be sterilized by conventional methods. The compact Swivel Connector may be rotated a full 360 degrees affording maximum positioning and flexibility. For use on all endotracheal tubes.