Learn More
Med Vet International Endotracheal Tube, J0148S, Red Rubber, 12.0mm, Each
Catalog No. 5000583618
Encompass_Preferred
Available in .5mm increments from 2.5 to 16.0mm. Murphy eye. Red rubber. Hard-to-find extra-large size now available. (adaptor not included except where specified)
Catalog No. 50-005-83618 Supplier Med Vet International Supplier No. JOR148S
Available in .5mm increments from 2.5 to 16.0mm. Murphy eye. Red rubber. Hard-to-find extra-large size now available. (adaptor not included except where specified)