Learn More
ILEX LIFE SCIENCES LLC EndoTrap Empty Columns Small (for 0.2-2 mL resin), pack of 5, endotoxin-removal accessory
SDP
Catalog No. 502339591
Encompass_Preferred
Empty columns (small size) for use with EndoTrap endotoxin (LPS) removal products. This column size is suitable for use with 0.2-2 mL resin. Pack of 5 columns. EndoTrap is an affinity chromatography resin based on a bacteriophage-derived protein which binds to endotoxins with high affinity and specificity. The EndoTrap protein captures the conserved region of the inner core of LPS molecules and is thereby able to bind to all kind of endotoxins from Gram-negative bacteria. It is neither an antibody, a synthetic peptide nor Polymyxin B-based. The EndoTrap family consists of the EndoTrap HD and EndoTrap red product lines.
Catalog No. 50-233-9591 Supplier ILEX LIFE SCIENCES LLC Supplier No. LET0020
Empty columns (small size) for use with EndoTrap endotoxin (LPS) removal products. This column size is suitable for use with 0.2-2 mL resin. Pack of 5 columns. EndoTrap is an affinity chromatography resin based on a bacteriophage-derived protein which binds to endotoxins with high affinity and specificity. The EndoTrap protein captures the conserved region of the inner core of LPS molecules and is thereby able to bind to all kind of endotoxins from Gram-negative bacteria. It is neither an antibody, a synthetic peptide nor Polymyxin B-based. The EndoTrap family consists of the EndoTrap HD and EndoTrap red product lines.