Learn More
BioVendor, LLC EndoTrap HD 1/1 Endotoxin Removal Kit. Includes: 1 x 1 ml column, 125 ml 5x Equilibration Buffer, 125 ml 5x Regeneration Buffer, 125 ml 5x Storage Buffer
SDP
Supplier:  BioVendor, LLC LET0009
Encompass
EndoTrap HD 1/1 Endotoxin Removal Kit. Includes: 1 x 1 ml column, 125 ml 5x Equilibration Buffer, 125 ml 5x Regeneration Buffer, 125 ml 5x Storage Buffer
Catalog No. 50-165-5481