Learn More
BioVendor, LLC EndoTrap HD 5x Equilibration Buffer, 5x concentrated, 125 ml
SDP
Supplier:  BioVendor, LLC LET0015
Encompass
EndoTrap HD 5x Equilibration Buffer, 5x concentrated, 125 ml. Component for EndoTrap HD. EndoTrap HD removes endotoxins (lipopolysaccharides/LPS) from aqueous solutions of samples such as proteins, antibodies, vaccines, nucleic acids, buffers and cell extracts with excellent sample recovery rate (>95%). EndoTrap HD is designed to meet the needs of pharma/biotech industry for efficient removal of LPS in biomanufacturing processes. EndoTrap HD fulfils all requirements for industrial applications, and can easily be integrated in early or late-stage purification steps. Manufactured by LIONEX GmbH in Germany. LPS binding capacity > 5,000,000 EU/ml. Removes >99.99% of LPS. Non-toxic bacteriophage-derived ligand, no cytotoxic effects. Re-use >10 times. Sample pH range 4-10. Suitable for high salt conditions. Suitable for TRIS, HEPES, MOPS or similar buffered samples. PBS buffer possible when enriched freshly with 0.1 mM calcium (magnesium) directly before use.
Catalog No. 50-206-9081