Learn More
ILEX LIFE SCIENCES LLC LIONEX EndoTrap red 5 ml settled resin Endotoxin Removal Resin 5 ML 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC LET0033
Encompass_Preferred
EndoTrap red 5 5 mL settled resin Affinity chromatography endotoxin (LPS) removal resin EndoTrap red endotoxin removal products manufactured in Germany by LIONEX are intended for removal of lipopolysaccharide (LPS) from biological samples in aqueous solutions such as proteins antibodies cell extracts and nucleic acids EndoTrap red is specially developed for buffers containing calcium chelators and for low-salt conditions EndoTrap red works effectively in the pH range of 6-9 and NaCl concentrations in the range of 50-250 mM Buffers such as HEPES PBS TRIS MOPS MES PIPES and also Citrate Acetate Glycine and Carbonate buffers are recommended EndoTrap red can be used with chelators of divalent cations (like EDTA) Phosphate buffers with NaCl concentrations below 100 mM are recommended for EndoTrap red LPS Binding Capacity 2 000 000 EU/ml Sample Recovery Rate 95% pH Sample Range 6-9 Non-toxic no cytotoxic effects Re-use at least 3 times
Catalog No. 50-212-0026