Learn More
Aviva Systems Biology Corp Endou Antibody - middle region (ARP42275_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42275_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Endou. It was validated on Western Blot. . Alternate names: Pp11r, Tcl-30ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-989