Learn More
Medchemexpress LLC HY-105917 10mg Medchemexpress, Endovion CAS:265646-85-3 Purity:>98%
SDP
Catalog No. 501968302
Encompass_Preferred
Medchemexpress, HY-105917 10mg Endovion CAS:265646-85-3 Endovion (NS3728) is a pharmacological anion channel inhibitor (like chloride channel) and the specific VRAC/VSOAC blocker. Endovion (NS3728) is also an Anoctamin-1 (ANO 1) channel inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8302 Supplier Medchemexpress LLC Supplier No. HY10591710MG
Medchemexpress, HY-105917 10mg Endovion CAS:265646-85-3 Endovion (NS3728) is a pharmacological anion channel inhibitor (like chloride channel) and the specific VRAC/VSOAC blocker. Endovion (NS3728) is also an Anoctamin-1 (ANO 1) channel inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.