Learn More
Medchemexpress LLC Endoxifen (E-isomer) | 114828-90-9 | 98.0% | 373.49 | 100 MG
SDP
Catalog No. 5000388019
Encompass_Preferred
Endoxifen E-isomer (E-Endoxifen) is an impurity in Endoxifen Z-isomer, exhibiting antiestrogenic effects. It is intended for research use only.
  • Inhibits PGR gene expression in E2-induced MCF-7 cells
  • Suitable for in vitro biological activity studies
  • Provided as a solid for various experimental applications
Catalog No. 50-003-88019 Supplier Medchemexpress LLC Supplier No. HY18719D100MG
Endoxifen E-isomer (E-Endoxifen) is an impurity in Endoxifen Z-isomer, exhibiting antiestrogenic effects. It is intended for research use only.
  • Inhibits PGR gene expression in E2-induced MCF-7 cells
  • Suitable for in vitro biological activity studies
  • Provided as a solid for various experimental applications