Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Endoxifen (E-isomer) | 114828-90-9 | MFCD09840374 | 98.0% | 373.49 g/mol | C25H27NO2 | 10 MG
SDP

Catalog No. 5000294815
Encompass_Preferred

Endoxifen (E-isomer) is the E-isomer of endoxifen provided for research use. It is reported as an impurity in the Z-isomer and exhibits antiestrogenic activity; intended for laboratory research and not for human use.

  • Exhibits antiestrogenic effects.
  • Purity 98.02%.
  • Chemical formula C25H27NO2; molecular weight 373.49 g/mol.
  • Soluble in DMSO at 50 mg/mL; may require ultrasonic treatment.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (2 years).

Catalog No. 50-002-94815 Supplier Medchemexpress LLC Supplier No. HY18719D10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Endoxifen (E-isomer) is the E-isomer of endoxifen provided for research use. It is reported as an impurity in the Z-isomer and exhibits antiestrogenic activity; intended for laboratory research and not for human use.

  • Exhibits antiestrogenic effects.
  • Purity 98.02%.
  • Chemical formula C25H27NO2; molecular weight 373.49 g/mol.
  • Soluble in DMSO at 50 mg/mL; may require ultrasonic treatment.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.