Learn More
Selleck Chemical LLC Endoxifen HCl-10mg
SDP
Catalog No. 502389964
Encompass_Preferred
Endoxifen HCl, the active metabolite of Tamoxifen, ia a potent and selective estrogen receptor antagonist. Phase 2.
Catalog No. 50-238-9964 Supplier Selleck Chemical LLC Supplier No. S783910MG
Endoxifen HCl, the active metabolite of Tamoxifen, ia a potent and selective estrogen receptor antagonist. Phase 2.