Learn More
Medchemexpress LLC HY-18719B 10mg Medchemexpress, Endoxifen (hydrochloride) CAS:1197194-41-4 Purity:>98%
SDP
Catalog No. 502082077
Encompass_Preferred
Medchemexpress, HY-18719B 10mg Endoxifen (hydrochloride) CAS:1197194-41-4 Endoxifen hydrochloride is a key active metabolite of Tamoxifen (TAM) with higher affinity and specificity to estrogen receptor that also inhibits aromatase activity. Endoxifen hydrochloride has the potential for breast cancer study. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2077 Supplier Medchemexpress LLC Supplier No. HY18719B10MG
Medchemexpress, HY-18719B 10mg Endoxifen (hydrochloride) CAS:1197194-41-4 Endoxifen hydrochloride is a key active metabolite of Tamoxifen (TAM) with higher affinity and specificity to estrogen receptor that also inhibits aromatase activity. Endoxifen hydrochloride has the potential for breast cancer study. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.