Learn More
Medchemexpress LLC Endoxifen hydrochloride | 1197194-41-4 | 98.0% | 409.95 | 5 MG
SDP
Catalog No. 5000386062
Encompass_Preferred
Endoxifen hydrochloride is a key active metabolite of Tamoxifen (TAM) with higher affinity and specificity to the estrogen receptor, and it also inhibits aromatase activity. It has potential for breast cancer study.
  • Key active metabolite of Tamoxifen (TAM).
  • Higher affinity and specificity to estrogen receptor.
  • Inhibits aromatase activity.
  • Potential for breast cancer study.
  • Approximately 100-fold more potent as an antagonist of the ER than tamoxifen.
  • Targets estrogen receptor α for degradation in breast cancer cells.
  • Blocks ERA transcriptional activity and inhibits estrogen-induced breast cancer cell proliferation.
Catalog No. 50-003-86062 Supplier Medchemexpress LLC Supplier No. HY18719B5MG
Endoxifen hydrochloride is a key active metabolite of Tamoxifen (TAM) with higher affinity and specificity to the estrogen receptor, and it also inhibits aromatase activity. It has potential for breast cancer study.
  • Key active metabolite of Tamoxifen (TAM).
  • Higher affinity and specificity to estrogen receptor.
  • Inhibits aromatase activity.
  • Potential for breast cancer study.
  • Approximately 100-fold more potent as an antagonist of the ER than tamoxifen.
  • Targets estrogen receptor α for degradation in breast cancer cells.
  • Blocks ERA transcriptional activity and inhibits estrogen-induced breast cancer cell proliferation.