Learn More
Zymo Research Corporation EndoZero III Spin Column w/15 ml Reservoir-X
SDP
Catalog No. NC2144352
Encompass_Preferred
Available as a refill for the ZymoPURE II Plasmid Gigaprep Kit. The EndoZero III Spin-Column with 15 ml Reservoir-X features durable polypropylene construction and can be used with centrifuges for the removal of residual endotoxins in plasmid DNA prepared with the ZymoPURE II Plasmid Gigaprep Kit, which is the fastest and simplest method available to efficiently isolate up to 25 mg of endotoxin-free plasmid DNA from E. coli. Capacity with15 ml Reservoir-X is 15 ml.
This item is not returnable. View return policy
Catalog No. NC2144352 Supplier Zymo Research Corporation Supplier No. C10965
Available as a refill for the ZymoPURE II Plasmid Gigaprep Kit. The EndoZero III Spin-Column with 15 ml Reservoir-X features durable polypropylene construction and can be used with centrifuges for the removal of residual endotoxins in plasmid DNA prepared with the ZymoPURE II Plasmid Gigaprep Kit, which is the fastest and simplest method available to efficiently isolate up to 25 mg of endotoxin-free plasmid DNA from E. coli. Capacity with15 ml Reservoir-X is 15 ml.