Learn More
BioVendor, LLC ENDOZYME II RECOMB. FAC.C ENDO (192 TESTS)
SDP
Catalog No. NC1386139
Encompass
DescriptionThe ENDONEXT™ EndoZyme II assay is the enhanced second generation of EndoZyme, based on recombinant Factor C (rFC), the synthetic analogue to the native endotoxin-specific principal receptor in the Limulus Amebocyte Lysate (LAL) cascade. Instead of being based on an animal source, EndoZyme II is a sustainable alternative method in line with regulatory requirements. EndoZyme II is particularly suited for end-product testing and in-process control of water and raw materials, as well as medical device testing.ShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationUnopened kits are stable at 2–8°C until the expiry date printed on the label.Calibration Range0.005 – 50 EU/mlLimit of Detection0.005 EU/mL
Catalog No. NC1386139 Supplier BioVendor, LLC Supplier No. 890030
DescriptionThe ENDONEXT™ EndoZyme II assay is the enhanced second generation of EndoZyme, based on recombinant Factor C (rFC), the synthetic analogue to the native endotoxin-specific principal receptor in the Limulus Amebocyte Lysate (LAL) cascade. Instead of being based on an animal source, EndoZyme II is a sustainable alternative method in line with regulatory requirements. EndoZyme II is particularly suited for end-product testing and in-process control of water and raw materials, as well as medical device testing.ShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationUnopened kits are stable at 2–8°C until the expiry date printed on the label.Calibration Range0.005 – 50 EU/mlLimit of Detection0.005 EU/mL