Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Endrin ketone | 53494-70-5 | MFCD16660998 | 380.9 g/mol | C12H8Cl6O | 10 MG
SDP

Catalog No. 5000284289
Encompass_Preferred

Endrin ketone is a chlorinated degradation product of the insecticide endrin used as an analytical reference in laboratory research. It is intended for research use only and is not for human or clinical use. Handle with care and follow applicable safety and regulatory guidance because endrin and its degradation products are carcinogenic.

  • CAS number 53494-70-5.
  • Chemical formula C12H8Cl6O.
  • Molecular weight 380.9 g/mol.
  • Available in small-mass vials (10 mg typical) for analytical applications.
  • Intended for research use only; not for human use.
  • May be subject to regulatory restrictions in certain territories.
  • Certificate of analysis and purity documentation available from the supplier.

Catalog No. 50-002-84289 Supplier Medchemexpress LLC Supplier No. HYW58589310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Endrin ketone is a chlorinated degradation product of the insecticide endrin used as an analytical reference in laboratory research. It is intended for research use only and is not for human or clinical use. Handle with care and follow applicable safety and regulatory guidance because endrin and its degradation products are carcinogenic.

  • CAS number 53494-70-5.
  • Chemical formula C12H8Cl6O.
  • Molecular weight 380.9 g/mol.
  • Available in small-mass vials (10 mg typical) for analytical applications.
  • Intended for research use only; not for human use.
  • May be subject to regulatory restrictions in certain territories.
  • Certificate of analysis and purity documentation available from the supplier.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.