Learn More
Medchemexpress LLC Endrin ketone | 53494-70-5 | MFCD16660998 | 380.91 g/mol | C12H8Cl6O | 100 MG
SDP
Catalog No. 5000284106
Encompass_Preferred
Endrin ketone is an analytical standard and a degradation product of the insecticide endrin, provided for laboratory research and analytical applications. It appears as a stable white solid and should be handled according to its safety data sheet because endrin is a known carcinogen. Intended for research use only.
  • Analytical standard for method development and calibration.
  • Suitable for environmental and residue analysis.
  • Stable white solid physical form.
  • Molecular formula C12H8Cl6O; molecular weight 380.91 g/mol.
  • Lot-specific certificate of analysis available for purity information.
Catalog No. 50-002-84106 Supplier Medchemexpress LLC Supplier No. HYW585893100MG
Endrin ketone is an analytical standard and a degradation product of the insecticide endrin, provided for laboratory research and analytical applications. It appears as a stable white solid and should be handled according to its safety data sheet because endrin is a known carcinogen. Intended for research use only.
  • Analytical standard for method development and calibration.
  • Suitable for environmental and residue analysis.
  • Stable white solid physical form.
  • Molecular formula C12H8Cl6O; molecular weight 380.91 g/mol.
  • Lot-specific certificate of analysis available for purity information.