Learn More
Medchemexpress LLC Endrin ketone | 53494-70-5 | MFCD16660998 | 380.91 g/mol | C12H8Cl6O | 25 MG
SDP
Catalog No. 5000284110
Encompass_Preferred
Endrin ketone is an analytical standard and a degradation product of the organochlorine pesticide endrin. It is provided for research and analytical applications; endrin is classified as a carcinogen, so the compound should be handled with appropriate safety precautions. Certificates of analysis and safety documentation are referenced for lot-specific data.
Catalog No. 50-002-84110 Supplier Medchemexpress LLC Supplier No. HYW58589325MG
Endrin ketone is an analytical standard and a degradation product of the organochlorine pesticide endrin. It is provided for research and analytical applications; endrin is classified as a carcinogen, so the compound should be handled with appropriate safety precautions. Certificates of analysis and safety documentation are referenced for lot-specific data.