Learn More
Medchemexpress LLC Endrin ketone | 53494-70-5 | MFCD16660998 | ≥98.0% | 380.91 | C12H8Cl6O | 5 MG
SDP
Catalog No. 5000284636
Encompass_Preferred
Endrin ketone is the oxidative degradation product of endrin, provided as a research-grade chemical standard for analytical and degradation studies. The parent compound endrin is a known carcinogen; the product is intended for laboratory research only and requires appropriate safety measures.
  • Intended for research and analytical use.
  • High purity (≥98%).
  • Available in small, quantized masses for analytical workflows.
  • Supplied with documentation and purity certificates.
  • Handle with appropriate safety precautions due to related carcinogenic hazards.
Catalog No. 50-002-84636 Supplier Medchemexpress LLC Supplier No. HYW5858935MG
Endrin ketone is the oxidative degradation product of endrin, provided as a research-grade chemical standard for analytical and degradation studies. The parent compound endrin is a known carcinogen; the product is intended for laboratory research only and requires appropriate safety measures.
  • Intended for research and analytical use.
  • High purity (≥98%).
  • Available in small, quantized masses for analytical workflows.
  • Supplied with documentation and purity certificates.
  • Handle with appropriate safety precautions due to related carcinogenic hazards.