Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Endura™ Oilbloc™ Arc-Flash Goat-Grain Leather Drivers Gloves

Catalog No. 17401281
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-401-281 3X-Large
17-401-277 Medium
17-401-278 Large
17-401-279 X-Large
17-401-280 2X-Large
17-401-276 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-401-281 Supplier Superior Glove™ Supplier No. 378GOBKL3X
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goatskin leather provides abrasion resistance, while Oilbloc™ treatment ensures oil and water resistance.

  • Goatskin leather has a high tensile strength for increased abrasion resistance 
  • Treated with Oilbloc™ for oil and water resistance
  • Aramid Fiber provides 360° cut protection 
  • Drivers style design slips on easily with an elastic back for a snug fit 
  • Arc flash tested for level 4 with an ATPV of 41 cal/cm2

Specifications

Arc Flash Level 4
Certifications/Compliance ANSI/ISEA 105 A4, ANSI 360° Cut Protection, Arc Flash Category 4 (41 cal/cm2), ANSI Heat Level 3, ASTM D6413
Color Brown
Cut Resistance Rating ANSI A4
Hand Specific
Material Grain Goatskin Leather
Product Type Driver Gloves
Size 3X-Large
Style Driver
For Use With (Application) Automotive Assembly, Construction, Manufacturing, and Utility Work
Gender Men's
Glove Cuff Style Elasticated
Liner Material Aramid Fiber
Puncture Rating ANSI/ISEA 105-16 Level 4
Series 378GOBKL
Thumb Style Keystone
Product Line Endura
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.