Learn More
LGC GENOMICS LLC (Lucigen) Endura Chemically Competent Cells, 24 Rxns (DUOs)Clone and maintain unstable or repetitive sequences and lentiviral libraries with high efficiency. Ideal for GeCKO CRISPR library transformation.
Catalog No. NC2160374
Encompass
Clone repetitive sequences and lentiviral librariesStabilize direct repeats and create lentiviral constructsGenerate CRISPR-GeCKO-Libraries-Endura lentiviral guide RNA librariesRecommended in CRISPR GeCKO library protocolsChoose electrocompetent or chemically competent cellsHighest efficiency commercially available cells for lentiviral cloningEndura™ Competent Cells include Control DNA and Recovery Medium, and are packaged as SOLOs (1 transformation per tube) or DUOs (2 transformations per tube) as indicated. Endura Electrocompetent cells are provided in DUOs format only. Recovery Medium is also available separately. The specified transformation efficiencies are with pUC DNA, unless indicated otherwise.
This item is not returnable. View return policy
Catalog No. NC2160374 Supplier LGC GENOMICS LLC Supplier No. 602402
Clone repetitive sequences and lentiviral librariesStabilize direct repeats and create lentiviral constructsGenerate CRISPR-GeCKO-Libraries-Endura lentiviral guide RNA librariesRecommended in CRISPR GeCKO library protocolsChoose electrocompetent or chemically competent cellsHighest efficiency commercially available cells for lentiviral cloningEndura™ Competent Cells include Control DNA and Recovery Medium, and are packaged as SOLOs (1 transformation per tube) or DUOs (2 transformations per tube) as indicated. Endura Electrocompetent cells are provided in DUOs format only. Recovery Medium is also available separately. The specified transformation efficiencies are with pUC DNA, unless indicated otherwise.