Learn More
LGC GENOMICS LLC (Lucigen) Endura Electrocompetent Cells, 24 Rxns (DUOs)Clone and maintain unstable or repetitive sequences and lentiviral libraries with high efficiency. Ideal for GeCKO CRISPR library transformation.