Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Endura™ Arc-Flash Grain Goatskin Leather Drivers Gloves

Catalog No. 17489674
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-489-674 2X-Large
17-489-670 Small
17-489-671 Medium
17-489-672 Large
17-489-673 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-489-674 Supplier Superior Glove™ Supplier No. 378GKTFGXX
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High tensile strength grain goatskin leather provides abrasion resistance, with a Aramid Fiber liner for ANSI A4 cut protection.

  • Grain goatskin leather has a high tensile strength for abrasion resistance
  • Aramid Fiber provides 360° cut protection
  • Cut and sewn liner ensures a no-bunch custom fit
  • Keystone thumb enhances durability and comfort
  • Arc flash level 3 protection, with an Arc Thermal Performance Value (ATPV) of 36 cal/cm2

Specifications

Arc Flash Level 3
Certifications/Compliance ANSI/ISEA 105-2016 Cut A5, ANSI 360° Cut Protection, Arc Flash Category 3 (33 cal/cm2), ANSI Heat Level 3
Construction Cut and Sewn
Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A4
Hand Specific
Material Grain Goatskin Leather
Product Type Driver Gloves
Size 2X-Large
Style Driver
For Use With (Application) Appliance Manufacturing, Construction, General Maintenance, and Steel Plants
Gender Unisex
Glove Cuff Style Elasticated
Liner Material Aramid Fiber
Puncture Rating ANSI/ISEA 105-16 Level 3
Series 378GKTFG
Thumb Style Keystone
Product Line Endura
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.