Learn More
Grainger CORTECH Wall-Mount Desk, Polyethylene, 36 in. x 24 in., 1 ea.
Catalog No. 502779297
Encompass_Preferred
The Wall-Mount Desk is a one-piece, rotationally molded institutional desk designed for high-use, behavioral health, and corrections environments. This durable, ligature-resistant unit is constructed from high-impact polyethylene and provides a secure, easy-to-clean workspace that bolts to the wall via concealed pilot holes.
  • Rotationally molded polyethylene design offers high strength and durability.
  • Features a 5-degree slope on the writing surface.
  • Passes CAL 133 flammability standards.
  • Optional high-pressure laminate surface available for a tailored aesthetic.
  • Secure wall-mount design with concealed pilot holes.
  • Manufactured to ISO quality standards in the USA.
  • Easy to clean, sanitize, and maintain.
  • 300 lb weight capacity.
Catalog No. 50-277-9297 Supplier Grainger Supplier No. 19N176
The Wall-Mount Desk is a one-piece, rotationally molded institutional desk designed for high-use, behavioral health, and corrections environments. This durable, ligature-resistant unit is constructed from high-impact polyethylene and provides a secure, easy-to-clean workspace that bolts to the wall via concealed pilot holes.
  • Rotationally molded polyethylene design offers high strength and durability.
  • Features a 5-degree slope on the writing surface.
  • Passes CAL 133 flammability standards.
  • Optional high-pressure laminate surface available for a tailored aesthetic.
  • Secure wall-mount design with concealed pilot holes.
  • Manufactured to ISO quality standards in the USA.
  • Easy to clean, sanitize, and maintain.
  • 300 lb weight capacity.