Learn More
Pacer Global LLC Endurance Pump Tubing, Size 16, Platinum Cured Silicone, 0.125" ID x 0.250" OD, 50-Foot Coil, 1/EA
SDP
Catalog No. 5000747444
Encompass_Preferred
Endurance Pump Tubing, Size 16, Platinum Cured Silicone, 0.125" ID x 0.250" OD, 50-Foot Coil, 1/EA
Catalog No. 50-007-47444 Supplier Pacer Global LLC Supplier No. MLSENDURAP0125025050
Endurance Pump Tubing, Size 16, Platinum Cured Silicone, 0.125" ID x 0.250" OD, 50-Foot Coil, 1/EA