Learn More
Pacer Global LLC Endurance Pump Tubing, Size 16, Platinum Cured Silicone, 0.125" ID x 0.250" OD, 500-Foot Spool, 1/EA
SDP
Catalog No. 5000747445
Encompass_Preferred
Endurance Pump Tubing, Size 16, Platinum Cured Silicone, 0.125" ID x 0.250" OD, 500-Foot Spool, 1/EA
Catalog No. 50-007-47445 Supplier Pacer Global LLC Supplier No. ENDURAP01250250500
Endurance Pump Tubing, Size 16, Platinum Cured Silicone, 0.125" ID x 0.250" OD, 500-Foot Spool, 1/EA