Learn More
Pacer Global LLC Endurance Pump Tubing, Size 17, Platinum Cured Silicone, 0.250" ID x 0.375" OD, 500-Foot Spool, 1/EA
SDP
Catalog No. 5000747441
Encompass_Preferred
Endurance Pump Tubing, Size 17, Platinum Cured Silicone, 0.250" ID x 0.375" OD, 500-Foot Spool, 1/EA
Catalog No. 50-007-47441 Supplier Pacer Global LLC Supplier No. ENDURAP02500375500
Endurance Pump Tubing, Size 17, Platinum Cured Silicone, 0.250" ID x 0.375" OD, 500-Foot Spool, 1/EA