Learn More
Cole-Parmer Enduro E0160 Gel XL Electrophoresis System 120V
Supplier:  Cole-Parmer 1731411
Encompass_Preferred
This convenient, complete system offers flexible run options and built-in safety features. Directly connect power supply to the gel boxeliminating the need for power leads. Large digital display and easy-to-use keypad provide simple operation and programming.
Catalog No. NC0162089