Learn More
Med Vet International Enema/Oral Calf Feeder Bag, J0139, 3000mL, Each
Catalog No. 5000581044
Encompass_Preferred
This bag was originally developed for enemas on larger patients. It has also found use as an oral calf feeder. Plus, it is less expensive. Short rigid end probe: helps ease passage. Round probe end with side port: helps prevent occlusion.
Catalog No. 50-005-81044 Supplier Med Vet International Supplier No. JOR139
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This bag was originally developed for enemas on larger patients. It has also found use as an oral calf feeder. Plus, it is less expensive. Short rigid end probe: helps ease passage. Round probe end with side port: helps prevent occlusion.