Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Energized Electrical Work Permit

Catalog No. 19109754 Shop All Brady Corporation Products
Change view
Click to view available options
Quantity:
25/Pad
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-109-754 25/Pad
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-109-754 Supplier Brady™ Supplier No. 99287

missing translation for 'validMainframeMsg'

25 permits per pad. The Brady™ Energized Electrical Work Permit meets NFPA 70E 130.1(A) requirements.

  • Meets NFPA 70E 130.1(A) requirements which mandate the use of a written permit when performing live electrical work
  • Made of 2-ply carbonless copy paper
  • Sold in packages of 25

Specifications

Width (English) Page 8.5 in.
Length (English) Page 11 in.
Color Black on White
Dimensions (L x W) 8.5 x 11 in.
Product Type 2-ply Carbonless
Quantity 25/Pad
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.